Learn More
Bioworld ACES (N-[carbamoylmethyl]-2-Aminoethane-sulfonic acid) Buffer, 0.5M, pH 6.2, 100 mL
Catalog No. 50255310
Encompass_Preferred
Synonym:N-(2-Acetamido)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)taurineACES is one ofGood's buffersdeveloped in the 1960s to provide buffers withpHranging from 6.15-8.35 for use in various applications. Azwitterionicbuffer with apKaof 6.9, it is often used as abuffering agentinbiologicalandbiochemicalresearch. ACES had been used to develop buffers for bothagaroseandpolyacrylamide gel electrophoresis.A saturated solution contains 0.22 M at 0°C. ACES binds Cu2+, but not significantly Mg2+, Ca2+or Mn2+.Appearance: LiquidHS Code : 29241900Melting Point: >220 °C (dec.)(lit.)PH Value: 6.0
Catalog No. 50-255-310 Supplier Bioworld Supplier No. 401200011
Synonym:N-(2-Acetamido)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)taurineACES is one ofGood's buffersdeveloped in the 1960s to provide buffers withpHranging from 6.15-8.35 for use in various applications. Azwitterionicbuffer with apKaof 6.9, it is often used as abuffering agentinbiologicalandbiochemicalresearch. ACES had been used to develop buffers for bothagaroseandpolyacrylamide gel electrophoresis.A saturated solution contains 0.22 M at 0°C. ACES binds Cu2+, but not significantly Mg2+, Ca2+or Mn2+.Appearance: LiquidHS Code : 29241900Melting Point: >220 °C (dec.)(lit.)PH Value: 6.0