Learn More
Bioworld ACES (N-[carbamoylmethyl]-2-Aminoethane-sulfonic acid) Buffer, 0.5M, pH 6.2, 250 mL
Catalog No. 50255311
Encompass_Preferred
Synonym:N-(2-Acetamido)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)taurineACES is one ofGood's buffersdeveloped in the 1960s to provide buffers withpHranging from 6.15-8.35 for use in various applications. Azwitterionicbuffer with apKaof 6.9, it is often used as abuffering agentinbiologicalandbiochemicalresearch. ACES had been used to develop buffers for bothagaroseandpolyacrylamide gel electrophoresis.A saturated solution contains 0.22 M at 0°C. ACES binds Cu2+, but not significantly Mg2+, Ca2+or Mn2+.Appearance: LiquidHS Code : 29241900Melting Point: >220 °C (dec.)(lit.)PH Value: 6.0
Catalog No. 50-255-311 Supplier Bioworld Supplier No. 401200012
Synonym:N-(2-Acetamido)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)-2-aminoethanesulfonic acid; N-(Carbamoylmethyl)taurineACES is one ofGood's buffersdeveloped in the 1960s to provide buffers withpHranging from 6.15-8.35 for use in various applications. Azwitterionicbuffer with apKaof 6.9, it is often used as abuffering agentinbiologicalandbiochemicalresearch. ACES had been used to develop buffers for bothagaroseandpolyacrylamide gel electrophoresis.A saturated solution contains 0.22 M at 0°C. ACES binds Cu2+, but not significantly Mg2+, Ca2+or Mn2+.Appearance: LiquidHS Code : 29241900Melting Point: >220 °C (dec.)(lit.)PH Value: 6.0