Learn More
Research Products International Corp ACES [N-(2-Acetamido)-2-aminoethane], 100 Grams
SDP
Catalog No. 501368060
Encompass_Preferred
Zwitterionic buffer used in biochemistry and molecular biology research.
  • pKa (25°C): 6.88
  • The working pH range in aqueous solution is 6.1 - 7. 5
  • Absorbance @ 280nm (5% water): <0.02
Catalog No. 50-136-8060 Supplier Research Products International Corp Supplier No. A22500100.0
Zwitterionic buffer used in biochemistry and molecular biology research.
  • pKa (25°C): 6.88
  • The working pH range in aqueous solution is 6.1 - 7. 5
  • Absorbance @ 280nm (5% water): <0.02