Learn More
Research Products International Corp ACES, 5 KG
SDP
Catalog No. 501976861
Encompass_Preferred
Zwitterionic buffer used in biochemistry and molecular biology research.
  • pKa (25°C): 6.88
  • The working pH range in aqueous solution is 6.1 - 7. 5
  • Absorbance @ 280nm (5% water): <0.02
Catalog No. 50-197-6861 Supplier Research Products International Corp Supplier No. A225005000.0
Add to Cart
Add to Cart
Zwitterionic buffer used in biochemistry and molecular biology research.
  • pKa (25°C): 6.88
  • The working pH range in aqueous solution is 6.1 - 7. 5
  • Absorbance @ 280nm (5% water): <0.02