Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals ACES, 99%, extra pure

Catalog No. AC228001000
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC228001000 100 g
AC228000250 25 g
AC228002500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228001000 Supplier Thermo Scientific Chemicals Supplier No. 228001000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N

Specifications

Absorbance (c=5, H2O), (c=5, H2O) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
Absorbance Conditions (c=5, H2O)
CAS Min % 98.5
Color White
CAS Max % 100.0
pH 3.6 to 4.4 (1% soln.)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula H2NCOCH2NHCH2CH2SO3H
Merck Index 15, 37
Formula Weight 182.19
Percent Purity ≥98.5%
Grade Extra Pure
Chemical Name or Material ACES, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.