Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACES, Biological Buffer, 98%, Spectrum™ Chemical
SDP

Catalog No. 18608314 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-314 25 g Amber Glass Bottle
18-608-312 100 g Amber Glass Bottle
18-608-311 1 kg Amber Glass Bottle
18-608-313 12 kg Amber Glass Bottle
18-608-315 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-314 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AC09025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Specifications

CAS Max % 100%
For Use With (Application) Molecular Biology
Physical Form Crystalline Powder
Quantity 25 g
Loss on Drying 0.2%
Packaging Amber Glass Bottle
Percent Purity 98%
Grade Reagent
Chemical Name or Material N-(2-Acetamido)-2-Aminoethanesulfonic Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.