Learn More
Bioworld ACES, 100 g
Catalog No. 50255184
Encompass_Preferred
ACES, High Purity Grade N-[carbamoylmethyl]-2-Aminoethane-sulfonic acidpKa: (20°C): 6.8+0.1Appearance: White crystalsHS Code : 29241900Melting Point: >220 °C (dec.)(lit.)PH Value: 6.1-7.5
Catalog No. 50-255-184 Supplier Bioworld Supplier No. 401000041
ACES, High Purity Grade N-[carbamoylmethyl]-2-Aminoethane-sulfonic acidpKa: (20°C): 6.8+0.1Appearance: White crystalsHS Code : 29241900Melting Point: >220 °C (dec.)(lit.)PH Value: 6.1-7.5