Learn More
Cayman Chemical Acesulfame-d4 (potassium salt), Purity: ≥99% deuterated forms (d1-d4), CAS: 1623054-53-4, 1 mg
SDP
Catalog No. 502941967
Encompass_Preferred
An internal standard for the quantification of acesulfame by GC- or LC-MS
Catalog No. 50-294-1967 Supplier Cayman Chemical Supplier No. 277861 MG
An internal standard for the quantification of acesulfame by GC- or LC-MS