Learn More
Medchemexpress LLC Acesulfame potassium | 55589-62-3 | 99.38% | 5 G
SDP
Catalog No. 5000410976
Encompass_Preferred
Acesulfame potassium is a synthetic sweetener that can affect cognitive function and may be related to immune evasion in cancer cells by suppressing autophagic degradation of PD-L1. It is for research use only and suitable for studies on neurological diseases, metabolic disorders, cancer, and immune evasion.
  • Upregulates PD-L1 protein levels in RIL-175 and SK-Hep1 cells.
  • Increases granzyme B production in RIL-175 and SK-Hep1 cells co-cultured with T cells.
  • Activates ERK1/2-mTORC1-ULK1 pathway to suppress autophagic degradation of PD-L1.
  • Inhibits mitochondrial metabolism and ATP production in SH-SY5Y cells.
  • Impairs energy metabolism and neuroprotective functions.
  • Increases body weight in male CD-1 mice and alters gut microbiome.
  • Affects neurometabolic functions in C57BL/6J mice with long-term intake.
Catalog No. 50-004-10976 Supplier Medchemexpress LLC Supplier No. HYD01955G
Acesulfame potassium is a synthetic sweetener that can affect cognitive function and may be related to immune evasion in cancer cells by suppressing autophagic degradation of PD-L1. It is for research use only and suitable for studies on neurological diseases, metabolic disorders, cancer, and immune evasion.
  • Upregulates PD-L1 protein levels in RIL-175 and SK-Hep1 cells.
  • Increases granzyme B production in RIL-175 and SK-Hep1 cells co-cultured with T cells.
  • Activates ERK1/2-mTORC1-ULK1 pathway to suppress autophagic degradation of PD-L1.
  • Inhibits mitochondrial metabolism and ATP production in SH-SY5Y cells.
  • Impairs energy metabolism and neuroprotective functions.
  • Increases body weight in male CD-1 mice and alters gut microbiome.
  • Affects neurometabolic functions in C57BL/6J mice with long-term intake.