Learn More
Medchemexpress LLC Acesulfame potassium | 55589-62-3 | 99.89% | 500 MG
SDP
Catalog No. 5000410679
Encompass_Preferred
Acesulfame potassium is an analytical standard intended for research and analytical applications. It is an artificial sweetener that may affect cognitive functions by altering neuro-metabolic functions in mice.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Commonly applied in HPLC, GC, and MS.
  • Grade of analytical standard.
  • Reference standard assay.
Catalog No. 50-004-10679 Supplier Medchemexpress LLC Supplier No. HYD0195R500MG
Acesulfame potassium is an analytical standard intended for research and analytical applications. It is an artificial sweetener that may affect cognitive functions by altering neuro-metabolic functions in mice.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Commonly applied in HPLC, GC, and MS.
  • Grade of analytical standard.
  • Reference standard assay.