Learn More
Medchemexpress LLC Acesulfame potassium (standard) | 55589-62-3 | 99.9% | 100 MG
SDP
Catalog No. 5000409758
Encompass_Preferred
This product serves as an analytical standard for Acesulfame (potassium), designated for various research and analytical applications. It is recognized as an artificial sweetener. Research indicates that long-term exposure to this compound may impact cognitive functions, potentially through modifications in neuro-metabolic processes within mice. The compound is specifically graded as an analytical standard, acting as a reference standard for assay purposes.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Commonly utilized in techniques such as High-Performance Liquid Chromatography (HPLC), Gas Chromatography (GC), and Mass Spectrometry (MS).
Catalog No. 50-004-09758 Supplier Medchemexpress LLC Supplier No. HYD0195R100MG
This product serves as an analytical standard for Acesulfame (potassium), designated for various research and analytical applications. It is recognized as an artificial sweetener. Research indicates that long-term exposure to this compound may impact cognitive functions, potentially through modifications in neuro-metabolic processes within mice. The compound is specifically graded as an analytical standard, acting as a reference standard for assay purposes.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Commonly utilized in techniques such as High-Performance Liquid Chromatography (HPLC), Gas Chromatography (GC), and Mass Spectrometry (MS).