Learn More
Medchemexpress LLC Acesulfame potassium (Standard) | 55589-62-3 | MFCD00043833 | 99.9% | 50 MG
SDP
Catalog No. 5000410680
Encompass_Preferred
Acesulfame potassium (Standard) is the analytical standard of Acesulfame potassium, intended for research and analytical applications. This artificial sweetener has been observed to affect cognitive functions, potentially via altering neuro-metabolic functions in mice. It serves as a reference standard for assay and is of analytical grade.
  • Used as a reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC
  • Suitable for GC
  • Suitable for MS
Catalog No. 50-004-10680 Supplier Medchemexpress LLC Supplier No. HYD0195R50MG
Acesulfame potassium (Standard) is the analytical standard of Acesulfame potassium, intended for research and analytical applications. This artificial sweetener has been observed to affect cognitive functions, potentially via altering neuro-metabolic functions in mice. It serves as a reference standard for assay and is of analytical grade.
  • Used as a reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC
  • Suitable for GC
  • Suitable for MS