Learn More
Medchemexpress LLC Acetaldehyde 2,4-dinitrophenylhydrazone-3,5,6-d3 | 259824-51-6 | 99.0% | C8H5D3N4O4 | 1 MG
SDP
Catalog No. 5000353030
Encompass_Preferred
Acetaldehyde 2,4-Dinitrophenylhydrazone-3,5,6-d3 is a deuterium-labeled form of Acetaldehyde 2,4-Dinitrophenylhydrazone-3,5,6. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for use as tracers for quantitation during the drug development process. Deuteration is particularly noted for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis through NMR, GC-MS, or LC-MS.
Catalog No. 50-003-53030 Supplier Medchemexpress LLC Supplier No. HY141855S1MG
Acetaldehyde 2,4-Dinitrophenylhydrazone-3,5,6-d3 is a deuterium-labeled form of Acetaldehyde 2,4-Dinitrophenylhydrazone-3,5,6. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for use as tracers for quantitation during the drug development process. Deuteration is particularly noted for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis through NMR, GC-MS, or LC-MS.