Learn More
Medchemexpress LLC Acetaldehyde 2,4-dinitrophenylhydrazone-3,5,6-d3 | 259824-51-6 | MFCD12022172 | 99.0% | 227.19 g/mol | C8H5D3N4O4 | 5 MG
SDP
Catalog No. 5000221748
Encompass_Preferred
Acetaldehyde 2,4-dinitrophenylhydrazone-3,5,6-d3 is a deuterium-labeled reference standard used for accurate quantitation and method validation of acetaldehyde derivatives in analytical workflows. It is isotopically labeled (d3), has a molecular weight of 227.19 g/mol, and a reported purity of 99.0%.
  • Isotopically labeled (d3) analog for use as an internal standard.
  • High purity (99.0%) suitable for quantitative analysis.
  • Supplied as a neat standard in small mg quantities for trace-level applications.
  • Compatible with GC-MS and LC-MS analytical methods.
  • Simplifies method validation and improves quantitation accuracy.
Catalog No. 50-002-21748 Supplier Medchemexpress LLC Supplier No. HY141855S5MG
Acetaldehyde 2,4-dinitrophenylhydrazone-3,5,6-d3 is a deuterium-labeled reference standard used for accurate quantitation and method validation of acetaldehyde derivatives in analytical workflows. It is isotopically labeled (d3), has a molecular weight of 227.19 g/mol, and a reported purity of 99.0%.
  • Isotopically labeled (d3) analog for use as an internal standard.
  • High purity (99.0%) suitable for quantitative analysis.
  • Supplied as a neat standard in small mg quantities for trace-level applications.
  • Compatible with GC-MS and LC-MS analytical methods.
  • Simplifies method validation and improves quantitation accuracy.