Learn More
Medchemexpress LLC Acetaldehyde, 2-(triphenylphosphoranylidene)- | 2136-75-6 | 99.7% | C20H17OP | 100 G
SDP
Catalog No. 5000433941
Encompass_Preferred
2-(Triphenylphosphoranylidene)acetaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Used as a Wittig reagent
  • Serves as a pharmaceutical intermediate
  • Employed as a raw material in organic synthesis and agrochemicals
Catalog No. 50-004-33941 Supplier Medchemexpress LLC Supplier No. HY75884100G
2-(Triphenylphosphoranylidene)acetaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Used as a Wittig reagent
  • Serves as a pharmaceutical intermediate
  • Employed as a raw material in organic synthesis and agrochemicals