Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acetaldehyde

Catalog No. 00070500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
00-070-500ML 500 mL
00-070-1L 1 L
00-070-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 00-070-500ML Supplier Sigma Aldrich Supplier No. 00070500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acetaldehyde

Specifications

CAS 75-07-0
Melting Point -125°C
Density 0.785 g/mL at 25°C
Boiling Point 21°C
Molecular Formula C2H4O
Refractive Index n20/D 1.332; n20/D 1.332
Linear Formula CH3CHO
MDL Number MFCD00006991
Quantity 500 mL
Synonym Ethanal
Molecular Weight (g/mol) 44.05 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.