Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023034 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3034 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-102-3034 Supplier MilliporeSigma Supelco Supplier No. RTC00007520ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 75-07-0
Melting Point -125°C (literature)
Density 0.785 g/mL (at 25°C (literature))
Boiling Point 21°C (literature)
Flash Point -40°C (Not Applicable)
Molecular Formula C2H4O
Refractive Index n20/D 1.332 (literature)
Linear Formula CH3CHO
Quantity 20 mL
MDL Number MFCD00006991
Synonym Acetaldehyde; Ethanal
Vapor Density 1.52 (vs air)
Vapor Pressure 14.63 psi (20°C)
Formula Weight 44.05
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.