Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde Semicarbazone 98.0+%, TCI America™
SDP

Catalog No. A01935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01935G 5 g
A019325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01935G Supplier TCI America Supplier No. A01935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 591-86-6
Molecular Formula C3H7N3O
Molecular Weight (g/mol) 101.109
MDL Number MFCD00059191
InChI Key IUNNCDSJWDQYPW-GORDUTHDSA-N
PubChem CID 5381348
IUPAC Name [(E)-ethylideneamino]urea
SMILES CC=NNC(=O)N

Specifications

Melting Point 164°C
Color White
Quantity 5 g
Formula Weight 101.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetaldehyde Semicarbazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.