Learn More
Chem Service Inc Acetaldehyde Solution,75-07-0,1ML
SDP
Supplier:  Chem Service Inc S11005A41ML
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Boiling Point 81.6 C; Melting Point -45 C; Composition: Acetonitrile 99.88%, Acetaldehyde 0.1%, Hydroquinone 0.02%
Catalog No. NC1379612