Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde Solution, Natural, 50 wt. in Ethanol, ≥98.0% (acetaldehyde & ethanol, GC), MilliporeSigma™ Supelco™

Catalog No. 111019322 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9322 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9322 Supplier MilliporeSigma Supelco Supplier No. 363925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetaldehyde comprises of a carbonyl group linked to a hydrogen atom. It is a well-known mutagen and carcinogen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00006991

Specifications

CAS 75-07-0
Density 0.868 g/mL (at 20°C)
Linear Formula CH3 CHO
Quantity 5 mL
Formula Weight 44.05
Percent Purity ≥98.0% (acetaldehyde & ethanol, GC)
Concentration or Composition (by Analyte or Components) ≥48.0% (acetaldehyde, T)
ShelfLife Limited shelf life, expiry date on the label
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.