Learn More
CPC Scientific Ac-Arg-Phe-Met-Trp-Met-Thr-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ACET001A
Encompass
Acetylated enkephalins, also named Opioid Receptor Antagonist, high affinity for m and K3 opioid receptor, weak affinity for K1 receptors and no affinity for K2 receptors.SEQUENCE: Ac-Arg-Phe-Met-Trp-Met-Thr-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Ac-RFMWMT-NH2MOLECULAR FORMULA: C42H61N11O8S2MOLECULAR WEIGHT: 912.15STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [152274-65-2]SYNONYMS: Opioid Receptor Antagonist 1 PeptideRESEARCH AREA: PainREFERENCES:C.T.Dooley et al., Proc. Natl. Acad. Sci. USA, 90, 10811 (1993)SKU(s): ACET-001A, ACET-001B
Catalog No. 50-210-8582