Learn More
Medchemexpress LLC Acetamide-2,2,2-d3 | 23724-60-9 | MFCD00190373 | 99.0% | 62.09 g/mol | C2H2D3NO | 10 MG
SDP
Catalog No. 5000282262
Encompass_Preferred
Acetamide-2,2,2-d3 is the trideuterated isotopologue of acetamide supplied for research use as an isotopic tracer and internal standard for quantitative analysis. It is used in NMR, GC-MS, and LC-MS workflows where deuterium labeling enables accurate tracking, quantitation, and method validation.
  • Deuterium-labeled (2,2,2-d3) isotopologue.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High isotopic enrichment (≈99 atom % D).
  • Provided in small analytical pack sizes (mg scale).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-82262 Supplier Medchemexpress LLC Supplier No. HYW24321710MG
Acetamide-2,2,2-d3 is the trideuterated isotopologue of acetamide supplied for research use as an isotopic tracer and internal standard for quantitative analysis. It is used in NMR, GC-MS, and LC-MS workflows where deuterium labeling enables accurate tracking, quantitation, and method validation.
  • Deuterium-labeled (2,2,2-d3) isotopologue.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High isotopic enrichment (≈99 atom % D).
  • Provided in small analytical pack sizes (mg scale).
  • For research use only; not for human or clinical use.