Learn More
Medchemexpress LLC Acetamide-2,2,2-d3, N-(4-hydroxyphenyl)- | 60902-28-5 | 100.0% | C8H6D3NO2 | 5 MG
SDP
Catalog No. 5000434266
Encompass_Preferred
Acetaminophen-d3 is the deuterium labeled form of Acetaminophen (Paracetamol), a widely used antipyretic and analgesic agent. It functions as a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM, and also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. This isotope-labeled compound is for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes for quantitation during drug development
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34266 Supplier Medchemexpress LLC Supplier No. HY66005S15MG
Acetaminophen-d3 is the deuterium labeled form of Acetaminophen (Paracetamol), a widely used antipyretic and analgesic agent. It functions as a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM, and also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. This isotope-labeled compound is for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes for quantitation during drug development
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs