Learn More
Medchemexpress LLC Acetamide-2,2,2-d3, N-(methyl-d3) | 3669-73-6 | MFCD01073934 | 98.6% | 79.13 g/mol | C3HD6NO | 1 MG
SDP
Catalog No. 5000299543
Encompass_Preferred
N-Methylacetamide-d6 is the deuterium-labeled analogue of N-methylacetamide provided as a high-purity stable isotope reagent for analytical and research applications. It is used as an internal standard and reference in mass spectrometry, NMR, and tracer studies.
  • High isotopic purity for accurate quantitation and spectral referencing.
  • Suitable as an internal standard for mass spectrometry analyses.
  • Provides a deuterium-labeled reference for NMR experiments.
  • Supplied in small, precise quantities for method development.
  • Certificate of analysis and safety data sheet available.
  • Molecular weight 79.13 g/mol, formula C3HD6NO.
Catalog No. 50-002-99543 Supplier Medchemexpress LLC Supplier No. HYW020018S1MG
N-Methylacetamide-d6 is the deuterium-labeled analogue of N-methylacetamide provided as a high-purity stable isotope reagent for analytical and research applications. It is used as an internal standard and reference in mass spectrometry, NMR, and tracer studies.
  • High isotopic purity for accurate quantitation and spectral referencing.
  • Suitable as an internal standard for mass spectrometry analyses.
  • Provides a deuterium-labeled reference for NMR experiments.
  • Supplied in small, precise quantities for method development.
  • Certificate of analysis and safety data sheet available.
  • Molecular weight 79.13 g/mol, formula C3HD6NO.