Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide-2,2,2-d3, N-(methyl-d3)- | 3669-73-6 | MFCD01073934 | 98.6% | 79.13 g/mol | C3HD6NO | 10 MG
SDP

Catalog No. 5000301476
Encompass_Preferred

N-Methylacetamide-d6 is the deuterium-labeled analogue of N-methylacetamide used as an isotopic standard and reagent for spectroscopic analysis, tracer studies, and synthetic applications. It is supplied as a colorless to light yellow liquid with high isotopic purity for reliable analytical results.

  • Deuterium labeled for use as an isotopic standard
  • High purity (98.6%) suitable for NMR and MS applications
  • Colorless to light yellow liquid for convenient handling
  • Molecular formula C3HD6NO; molecular weight 79.13 g/mol
  • CAS number 3669-73-6 for unambiguous identification
  • Storage at -20°C (pure) preserves stability for years

Catalog No. 50-003-01476 Supplier Medchemexpress LLC Supplier No. HYW020018S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methylacetamide-d6 is the deuterium-labeled analogue of N-methylacetamide used as an isotopic standard and reagent for spectroscopic analysis, tracer studies, and synthetic applications. It is supplied as a colorless to light yellow liquid with high isotopic purity for reliable analytical results.

  • Deuterium labeled for use as an isotopic standard
  • High purity (98.6%) suitable for NMR and MS applications
  • Colorless to light yellow liquid for convenient handling
  • Molecular formula C3HD6NO; molecular weight 79.13 g/mol
  • CAS number 3669-73-6 for unambiguous identification
  • Storage at -20°C (pure) preserves stability for years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.