Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide-2,2,2-d3, N-(methyl-d3)- | 3669-73-6 | MFCD01073934 | 98.6% | 79.13 g·mol⁻¹ | C3HD6NO | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW020018S

Encompass_Preferred

N-Methylacetamide-d6 is the deuterium-labeled analogue of N-methylacetamide supplied for research use as a stable isotope reference standard. It is a colorless liquid used as an internal standard or tracer in NMR, GC-MS, and LC-MS analyses, and for isotopic labeling studies, with high isotopic and chemical purity.

  • High chemical purity (98.6%).
  • Molecular formula C3HD6NO.
  • Molecular weight 79.13 g·mol⁻¹.
  • Available in small pack sizes, including 25 mg.
  • Recommended storage: pure form -20°C for long-term stability.
  • Suitable as an internal standard or tracer for NMR, GC-MS, and LC-MS.

Catalog No. 50-003-03802


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.