Learn More
Medchemexpress LLC Acetamide, 2-[[7-(3,4-dimethoxyphenyl)-2-quinoxalinyl)amino]-N-methyl- | 1513879-21-4 | 99.9% | 352.39 | 25 MG
SDP
Catalog No. 5000385930
Encompass_Preferred
BQR-695 is a PI4KIIIβ inhibitor with IC50s of 80 nM for human PI4KIIIβ and 3.5 nM for the *Plasmodium* variant of PI4KIIIβ. For research use only.
  • Treatment with 0.5 μM of either KAI407 or BQR695 causes GFP-PHOsh2 to redistribute to the parasite plasma membrane, consistent with depletion of intracellular PI4P upon inhibition of PfPI4K function.
  • BQR695 shows no evidence of toxicity against mature red blood cells (RBCs).
  • It induces a schizont-stage arrest indistinguishable from that observed in imidazopyrazine-treated parasites and exhibits cross-resistance with the imidazopyrazine-resistant lines.
  • DMSO: 50 mg/mL (141.89 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO).
  • Powder: -20°C for 3 years, 4°C for 2 years.
  • In solvent: -80°C for 2 years, -20°C for 1 year.
Catalog No. 50-003-85930 Supplier Medchemexpress LLC Supplier No. 25MGHY18748
BQR-695 is a PI4KIIIβ inhibitor with IC50s of 80 nM for human PI4KIIIβ and 3.5 nM for the *Plasmodium* variant of PI4KIIIβ. For research use only.
  • Treatment with 0.5 μM of either KAI407 or BQR695 causes GFP-PHOsh2 to redistribute to the parasite plasma membrane, consistent with depletion of intracellular PI4P upon inhibition of PfPI4K function.
  • BQR695 shows no evidence of toxicity against mature red blood cells (RBCs).
  • It induces a schizont-stage arrest indistinguishable from that observed in imidazopyrazine-treated parasites and exhibits cross-resistance with the imidazopyrazine-resistant lines.
  • DMSO: 50 mg/mL (141.89 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO).
  • Powder: -20°C for 3 years, 4°C for 2 years.
  • In solvent: -80°C for 2 years, -20°C for 1 year.