Learn More
Medchemexpress LLC Acetamide, 2-[[7-(3,4-dimethoxyphenyl)-2-quinoxalinyl)amino]-N-methyl- | 1513879-21-4 | 99.9% | 352.39 | 5 MG
SDP
Catalog No. 5000387916
Encompass_Preferred
BQR-695 is a PI4KIIIβ inhibitor, active against both human and *Plasmodium* variants of PI4KIIIβ. It effectively inhibits PI4KIIIβ function, leading to depletion of intracellular PI4P, and shows no toxicity against mature red blood cells. It induces schizont-stage arrest and exhibits cross-resistance with imidazopyrazine-resistant lines.
  • Inhibits human PI4KIIIβ with an IC50 of 80 nM
  • Inhibits *Plasmodium* PI4KIIIβ with an IC50 of 3.5 nM
  • Causes GFP-PHOsh2 to redistribute to parasite plasma membrane
  • No toxicity against mature red blood cells
  • Induces schizont-stage arrest
  • Exhibits cross-resistance with imidazopyrazine-resistant lines
Catalog No. 50-003-87916 Supplier Medchemexpress LLC Supplier No. HY187485MG
BQR-695 is a PI4KIIIβ inhibitor, active against both human and *Plasmodium* variants of PI4KIIIβ. It effectively inhibits PI4KIIIβ function, leading to depletion of intracellular PI4P, and shows no toxicity against mature red blood cells. It induces schizont-stage arrest and exhibits cross-resistance with imidazopyrazine-resistant lines.
  • Inhibits human PI4KIIIβ with an IC50 of 80 nM
  • Inhibits *Plasmodium* PI4KIIIβ with an IC50 of 3.5 nM
  • Causes GFP-PHOsh2 to redistribute to parasite plasma membrane
  • No toxicity against mature red blood cells
  • Induces schizont-stage arrest
  • Exhibits cross-resistance with imidazopyrazine-resistant lines