Learn More
Medchemexpress LLC Acetamide, 2-(aminooxy)-N-3,6,9,12-tetraoxapentadec-14-yn-1-yl- | 2253965-03-4 | 97.0% | 304.34 g/mol | C13H24N2O6 | 50 MG
SDP
Catalog No. 5000202404
Encompass_Preferred
Aminooxy-amido-PEG4-propargyl is a PEG4-based linker reagent used for bioconjugation and linker installation in research applications. It contains an aminooxy group and a terminal alkyne that enable orthogonal conjugation chemistry, and the PEG4 spacer improves solubility and flexibility for bioconjugates.
  • Contains aminooxy group for oxime ligation or conjugation.
  • Contains terminal alkyne (propargyl) for copper-catalyzed azide-alkyne cycloaddition.
  • PEG4 spacer increases solubility and reduces steric hindrance in conjugates.
  • High reported purity (97.0%) suitable for research applications.
  • Viscous, colorless to light yellow physical form for handling.
  • Available in small research pack size for lab-scale use.
Catalog No. 50-002-02404 Supplier Medchemexpress LLC Supplier No. HY13343550MG
Aminooxy-amido-PEG4-propargyl is a PEG4-based linker reagent used for bioconjugation and linker installation in research applications. It contains an aminooxy group and a terminal alkyne that enable orthogonal conjugation chemistry, and the PEG4 spacer improves solubility and flexibility for bioconjugates.
  • Contains aminooxy group for oxime ligation or conjugation.
  • Contains terminal alkyne (propargyl) for copper-catalyzed azide-alkyne cycloaddition.
  • PEG4 spacer increases solubility and reduces steric hindrance in conjugates.
  • High reported purity (97.0%) suitable for research applications.
  • Viscous, colorless to light yellow physical form for handling.
  • Available in small research pack size for lab-scale use.