Learn More
Medchemexpress LLC Acetamide | 60-35-5 | MFCD00008023 | 100.0% | 59.07 g/mol | C2H5NO | 500g
SDP
Catalog No. 5000266709
Encompass_Preferred
Acetamide is used as an intermediate in the synthesis of methylamine thioacetamide and insecticides and as a plasticizer in leather cloth and coatings Acetamide has carcinogenicity Acetamide derivatives may has antioxidant activity and potential anti-inflammatory activity Acetamide holds promise for research in the fields of anti-inflammatory and cancer studies[1][2][3]
Catalog No. 50-002-66709 Supplier Medchemexpress LLC Supplier No. HYY0946500G
Acetamide is used as an intermediate in the synthesis of methylamine thioacetamide and insecticides and as a plasticizer in leather cloth and coatings Acetamide has carcinogenicity Acetamide derivatives may has antioxidant activity and potential anti-inflammatory activity Acetamide holds promise for research in the fields of anti-inflammatory and cancer studies[1][2][3]