Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-[2-(4-hydroxyphenyl)ethyl]- | 1202-66-0 | 99.7% | 1 ML
SDP

Catalog No. 5000234400
Encompass_Preferred

N-Acetyltyramine is an acetylated derivative of tyramine used in research as a quorum-sensing inhibitor and a bacterial-targeted small molecule. It is supplied as a powder with guidance for preparing mM stock solutions and is soluble in DMSO for biochemical and microbiology assays.

  • CAS number 1202-66-0.
  • Molecular formula C10H13NO2.
  • Molecular weight 179.22 g/mol.
  • Purity 99.7% (HPLC).
  • Form powder suitable for preparing concentrated DMSO stocks.
  • Solubility in DMSO; solubility ≥ 5 mg/mL and typical DMSO stock up to 100 mg/mL.
  • Common use in quorum-sensing and bacterial assay research.

Catalog No. 50-002-34400 Supplier Medchemexpress LLC Supplier No. HY12050410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Acetyltyramine is an acetylated derivative of tyramine used in research as a quorum-sensing inhibitor and a bacterial-targeted small molecule. It is supplied as a powder with guidance for preparing mM stock solutions and is soluble in DMSO for biochemical and microbiology assays.

  • CAS number 1202-66-0.
  • Molecular formula C10H13NO2.
  • Molecular weight 179.22 g/mol.
  • Purity 99.7% (HPLC).
  • Form powder suitable for preparing concentrated DMSO stocks.
  • Solubility in DMSO; solubility ≥ 5 mg/mL and typical DMSO stock up to 100 mg/mL.
  • Common use in quorum-sensing and bacterial assay research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.