Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-[2-(methylthio)phenyl]- | 6310-41-4 | 99.0% | 181.25 | 50 MG
SDP

Catalog No. 5000387808
Encompass_Preferred

NSC-41589 is N-[2-(methylsulfanyl)phenyl]acetamide, intended for research use only. This product has not been fully validated for medical applications. It is a solid, white to off-white compound with high purity, offering reliable performance for various laboratory experiments.

  • High purity of 99.0%
  • Molecular weight of 181.25
  • Solid appearance, white to off-white
  • Soluble in DMSO up to 100 mg/mL
  • Stable for extended periods when stored properly as powder or in solvent

Catalog No. 50-003-87808 Supplier Medchemexpress LLC Supplier No. HY1859450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NSC-41589 is N-[2-(methylsulfanyl)phenyl]acetamide, intended for research use only. This product has not been fully validated for medical applications. It is a solid, white to off-white compound with high purity, offering reliable performance for various laboratory experiments.

  • High purity of 99.0%
  • Molecular weight of 181.25
  • Solid appearance, white to off-white
  • Soluble in DMSO up to 100 mg/mL
  • Stable for extended periods when stored properly as powder or in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.