Learn More
Medchemexpress LLC Acetamide, N-[3-(3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)phenyl] | 1401731-54-1 | 99.8% | 266.30 | C15H14N4O | 10 MG
SDP
Catalog No. 5000227056
Encompass_Preferred
A pyrazolo-pyridine analogue kinase inhibitor active against CK1 and CHK1, provided as a 10 mg research reagent with a molecular weight of 266.30 and formula C15H14N4O. It is supplied at high purity for use in biochemical and cellular studies.
  • Broad-spectrum kinase inhibition, including CK1δ and CHK1
  • Reported IC50 values: CK1δ 0.9 μM, Chk1 0.23 μM
  • High purity (99.8%) for reliable assay performance
  • Suitable for biochemical and cellular assays
  • Supplied in research-scale quantities for experimental work
Catalog No. 50-002-27056 Supplier Medchemexpress LLC Supplier No. HY11785710MG
A pyrazolo-pyridine analogue kinase inhibitor active against CK1 and CHK1, provided as a 10 mg research reagent with a molecular weight of 266.30 and formula C15H14N4O. It is supplied at high purity for use in biochemical and cellular studies.
  • Broad-spectrum kinase inhibition, including CK1δ and CHK1
  • Reported IC50 values: CK1δ 0.9 μM, Chk1 0.23 μM
  • High purity (99.8%) for reliable assay performance
  • Suitable for biochemical and cellular assays
  • Supplied in research-scale quantities for experimental work