Learn More
Medchemexpress LLC Acetamide, N-[3-(3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)phenyl]- | 1401731-54-1 | 99.8% | 266.30 | C15H14N4O | 100 MG
SDP
Catalog No. 5000227682
Encompass_Preferred
MRT00033659 (CAS 1401731-54-1) is a small-molecule kinase inhibitor reported to target CK1 and CHK1, supplied as a high-purity research reagent for in vitro and preclinical studies. The compound has formula C15H14N4O and a molecular weight of 266.30; it is unstable in solution and should be freshly prepared prior to use.
  • High purity suitable for analytical and biological studies.
  • Reported CK1 and CHK1 inhibitory activity for kinase research.
  • Unstable in solution-freshly prepare prior to experiments.
  • Available in small, research-scale quantities for assay work.
Catalog No. 50-002-27682 Supplier Medchemexpress LLC Supplier No. HY117857100MG
MRT00033659 (CAS 1401731-54-1) is a small-molecule kinase inhibitor reported to target CK1 and CHK1, supplied as a high-purity research reagent for in vitro and preclinical studies. The compound has formula C15H14N4O and a molecular weight of 266.30; it is unstable in solution and should be freshly prepared prior to use.
  • High purity suitable for analytical and biological studies.
  • Reported CK1 and CHK1 inhibitory activity for kinase research.
  • Unstable in solution-freshly prepare prior to experiments.
  • Available in small, research-scale quantities for assay work.