Learn More
Medchemexpress LLC Acetamide, N-[3-(3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)phenyl] | 1401731-54-1 | 99.8% | 266.30 g·mol⁻¹ | C15H14N4O | 50 MG
SDP
Catalog No. 5000227822
Encompass_Preferred
N-(3-(3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)phenyl)acetamide is a research-grade small-molecule inhibitor of casein kinase 1 (CK1) and checkpoint kinase 1 (CHK1), supplied as a solid for laboratory research use. It has CAS 1401731-54-1, molecular formula C15H14N4O, molecular weight 266.30 g·mol⁻¹, and reported purity of 99.77%.
  • High chemical purity suitable for biochemical and cellular assays.
  • Solid form enables straightforward handling and storage.
  • Targets CK1 and CHK1 for kinase pathway studies.
  • Available in small research quantities for assay development.
Catalog No. 50-002-27822 Supplier Medchemexpress LLC Supplier No. HY11785750MG
N-(3-(3-methyl-1H-pyrazolo[3,4-b]pyridin-5-yl)phenyl)acetamide is a research-grade small-molecule inhibitor of casein kinase 1 (CK1) and checkpoint kinase 1 (CHK1), supplied as a solid for laboratory research use. It has CAS 1401731-54-1, molecular formula C15H14N4O, molecular weight 266.30 g·mol⁻¹, and reported purity of 99.77%.
  • High chemical purity suitable for biochemical and cellular assays.
  • Solid form enables straightforward handling and storage.
  • Targets CK1 and CHK1 for kinase pathway studies.
  • Available in small research quantities for assay development.