Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-[4-(aminosulfonyl)phenyl]- | 121-61-9 | MFCD00035784 | 100.0% | 214.24 | C8H10N2O3S | 200 MG
SDP

Supplier:  Medchemexpress LLC HYW069721200MG

Encompass_Preferred

N-Acetylsulfanilamide is a research-grade small molecule reported to have potential anti-infective activity. It is supplied as a white to off-white solid and is commonly used as a reference standard or for laboratory research.

  • Cas number 121-61-9.
  • Molecular weight 214.24.
  • Purity 99.97% (manufacturer listed).
  • Appearance white to off-white solid.
  • Available pack sizes include 200 MG.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-10841


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.