Learn More
Medchemexpress LLC Acetamide, N-[4-(ethoxy-d5)phenyl]- | 69323-74-6 | 99.5% | 50 MG
SDP
Catalog No. 5000405524
Encompass_Preferred
Phenacetin-d5 is a deuterium-labeled form of Phenacetin, a non-opioid analgesic and antipyretic agent. It functions as a selective COX-3 inhibitor and is utilized as a probe for cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.
  • Deuterium-labeled Phenacetin
  • Non-opioid analgesic and antipyretic
  • Selective COX-3 inhibitor
  • Used as a probe for cytochrome P450 enzymes CYP1A2
Catalog No. 50-004-05524 Supplier Medchemexpress LLC Supplier No. HYB0476S50MG
Phenacetin-d5 is a deuterium-labeled form of Phenacetin, a non-opioid analgesic and antipyretic agent. It functions as a selective COX-3 inhibitor and is utilized as a probe for cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.
  • Deuterium-labeled Phenacetin
  • Non-opioid analgesic and antipyretic
  • Selective COX-3 inhibitor
  • Used as a probe for cytochrome P450 enzymes CYP1A2