Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-(4-formylphenyl)- | 122-85-0 | MFCD00003380 | 99.7% | 163.17 g/mol | C9H9NO2 | 50 G
SDP

Catalog No. 5000210738
Encompass_Preferred

N-(4-Formylphenyl)acetamide is an aromatic aldehyde used as a biochemical reagent and intermediate in organic synthesis. It is supplied as a white to yellow solid and is provided at high purity for laboratory research and assay development.

  • appearance: solid (white to yellow).
  • purity: 99.7%.
  • molecular weight: 163.17 g/mol.
  • Chemical formula: C9H9NO2.
  • cas number: 122-85-0.
  • storage: 4°C, stored under nitrogen.

Catalog No. 50-002-10738 Supplier Medchemexpress LLC Supplier No. HYW06744550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(4-Formylphenyl)acetamide is an aromatic aldehyde used as a biochemical reagent and intermediate in organic synthesis. It is supplied as a white to yellow solid and is provided at high purity for laboratory research and assay development.

  • appearance: solid (white to yellow).
  • purity: 99.7%.
  • molecular weight: 163.17 g/mol.
  • Chemical formula: C9H9NO2.
  • cas number: 122-85-0.
  • storage: 4°C, stored under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.