Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]- | 893990-34-6 | MFCD05957363 | 99.7% | 342.44 | C16H14N4OS2 | 10 MG
SDP

Catalog No. 5000289016
Encompass_Preferred

VU 0240551 is a small-molecule research compound that acts as a potent neuronal K+-Cl- cotransporter 2 (KCC2) inhibitor and is used in in vitro pharmacology and preclinical studies. It shows submicromolar activity at KCC2 and selectivity versus NKCC1.

  • Potent KCC2 inhibitor (IC50 ≈ 560 nM).
  • Selective versus NKCC1 (IC50 > 50 μM).
  • High purity (99.7%).
  • Molecular weight 342.44; formula C16H14N4OS2.
  • White to off-white solid with high solubility in DMSO (≥ 50 mg/mL).
  • Recommended storage: powder -20 °C (long-term) or 4 °C (shorter term); solvent storage at -80 °C.
  • Supplied in small pack sizes suitable for research (5 mg-100 mg).

Catalog No. 50-002-89016 Supplier Medchemexpress LLC Supplier No. HY1668910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU 0240551 is a small-molecule research compound that acts as a potent neuronal K+-Cl- cotransporter 2 (KCC2) inhibitor and is used in in vitro pharmacology and preclinical studies. It shows submicromolar activity at KCC2 and selectivity versus NKCC1.

  • Potent KCC2 inhibitor (IC50 ≈ 560 nM).
  • Selective versus NKCC1 (IC50 > 50 μM).
  • High purity (99.7%).
  • Molecular weight 342.44; formula C16H14N4OS2.
  • White to off-white solid with high solubility in DMSO (≥ 50 mg/mL).
  • Recommended storage: powder -20 °C (long-term) or 4 °C (shorter term); solvent storage at -80 °C.
  • Supplied in small pack sizes suitable for research (5 mg-100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.