Learn More
Medchemexpress LLC Acetamide, N-[(4S)-1-[(cyanoamino)[[3-(difluoromethoxy)phenyl]imino]methyl]-3-(3,4-dichlorophenyl)- | 1906919-67-2 | 99.3% | 525.34 | 25 MG
SDP
Catalog No. 5000403039
Encompass_Preferred
BAY-598 is a chemical probe and a selective small molecule inhibitor of SMYD2 with an IC50 of 27 nM. It is for research use only and not sold to patients.
  • Selective small molecule inhibitor of SMYD2 with an IC50 of 27 nM.
  • Blocks in vitro methylation of MAPKAPK3 by SMYD2 but has no activity against the SMYD2-related KMT SMYD3.
  • Reduces the growth of Kras;p53 mutant PDAC cells after 9 days in culture.
  • Solid appearance.
  • Light yellow to yellow color.
Catalog No. 50-004-03039 Supplier Medchemexpress LLC Supplier No. 25MGHY19546
BAY-598 is a chemical probe and a selective small molecule inhibitor of SMYD2 with an IC50 of 27 nM. It is for research use only and not sold to patients.
  • Selective small molecule inhibitor of SMYD2 with an IC50 of 27 nM.
  • Blocks in vitro methylation of MAPKAPK3 by SMYD2 but has no activity against the SMYD2-related KMT SMYD3.
  • Reduces the growth of Kras;p53 mutant PDAC cells after 9 days in culture.
  • Solid appearance.
  • Light yellow to yellow color.