Learn More
Medchemexpress LLC Acetamide, N-[[(5S)-3-[4-(methylsulfinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]- | 96800-41-8 | MFCD31382166 | 99.4% | 296.34 g/mol | C13H16N2O4S | 1 MG
SDP
Catalog No. 5000245258
Encompass_Preferred
DuP 105 is an orally active oxazolidinone-class antimicrobial research compound (CAS 96800-41-8) with reported activity against Gram-positive bacteria; it has been used in animal infection models and is intended for laboratory research use only.
  • Orally active oxazolidinone antimicrobial.
  • Demonstrated activity against Gram-positive bacteria, including staphylococcal and streptococcal strains.
  • Suitable for in vitro and in vivo studies of bacterial infection.
  • Provided as a white to off-white solid for laboratory use.
  • High purity, approximately 99.4% as supplied.
  • Stable with recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-45258 Supplier Medchemexpress LLC Supplier No. HY1017261MG
DuP 105 is an orally active oxazolidinone-class antimicrobial research compound (CAS 96800-41-8) with reported activity against Gram-positive bacteria; it has been used in animal infection models and is intended for laboratory research use only.
  • Orally active oxazolidinone antimicrobial.
  • Demonstrated activity against Gram-positive bacteria, including staphylococcal and streptococcal strains.
  • Suitable for in vitro and in vivo studies of bacterial infection.
  • Provided as a white to off-white solid for laboratory use.
  • High purity, approximately 99.4% as supplied.
  • Stable with recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).