Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-(6,9-dihydro-6-oxo-1H-purin-2-yl)- | 19962-37-9 | MFCD00078201 | 99.7% | 193.16 | C7H7N5O2 | 5 G
SDP

Catalog No. 5000209721
Encompass_Preferred

N2-Acetylguanine is a C2-modified guanine research chemical supplied as a high-purity powder for studies of nucleic acid interactions and riboswitch binding.

  • High purity: 99.69% (reported).
  • Molecular weight: 193.16 g/mol.
  • Chemical formula: C7H7N5O2.
  • Solubility: soluble in DMSO at 3.57 mg/mL with ultrasonic warming to 80°C.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available package: 5 g powder, suitable for laboratory research use.

Catalog No. 50-002-09721 Supplier Medchemexpress LLC Supplier No. HYW0407955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N2-Acetylguanine is a C2-modified guanine research chemical supplied as a high-purity powder for studies of nucleic acid interactions and riboswitch binding.

  • High purity: 99.69% (reported).
  • Molecular weight: 193.16 g/mol.
  • Chemical formula: C7H7N5O2.
  • Solubility: soluble in DMSO at 3.57 mg/mL with ultrasonic warming to 80°C.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available package: 5 g powder, suitable for laboratory research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.