Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-methyl-, N-d1- | 3669-70-3 | MFCD00008064 | 99.4% | 74.10 g/mol | C3H6DNO | 100 MG
SDP

Catalog No. 5000300268
Encompass_Preferred

N-Methylacetamide-d1 is a deuterium-labeled analogue of N-methylacetamide provided for laboratory research use. It is used as an isotope-labeled reference standard or tracer in analytical assays such as NMR, GC-MS, and LC-MS, and is supplied in small bench-scale pack sizes suitable for analytical workflows.

  • Deuterium-labeled at the nitrogen position.
  • High purity (99.4%) suitable for analytical applications.
  • Molecular weight 74.10 g/mol and formula C3H6DNO.
  • Available in small pack sizes, including 100 mg.
  • Store pure material at -20°C for long-term stability.
  • Intended for research use only as an analytical standard.

Catalog No. 50-003-00268 Supplier Medchemexpress LLC Supplier No. HYW020018S1100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methylacetamide-d1 is a deuterium-labeled analogue of N-methylacetamide provided for laboratory research use. It is used as an isotope-labeled reference standard or tracer in analytical assays such as NMR, GC-MS, and LC-MS, and is supplied in small bench-scale pack sizes suitable for analytical workflows.

  • Deuterium-labeled at the nitrogen position.
  • High purity (99.4%) suitable for analytical applications.
  • Molecular weight 74.10 g/mol and formula C3H6DNO.
  • Available in small pack sizes, including 100 mg.
  • Store pure material at -20°C for long-term stability.
  • Intended for research use only as an analytical standard.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.