Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetamide, N-(phenylmethyl)- | 588-46-5 | ≥97.0% | 149.19 | C9H11NO | 10 G
SDP

Catalog No. 5000209921
Encompass_Preferred

N-Benzylacetamide is an acetamide derivative supplied for research use and reported as an endogenous metabolite and a major component of the plasma metabolic profile of benznidazole. It is provided as a white to off-white solid and is characterized by a molecular formula of C9H11NO and a molecular weight of 149.19 g·mol⁻¹. Storage recommendations are 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light).

  • Cas number: 588-46-5.
  • Chemical formula: C9H11NO.
  • Molecular weight: 149.19 g·mol⁻¹.
  • Purity: ≥97.0%.
  • Appearance: white to off-white solid.
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light).
  • Synonyms: N-acetylbenzylamine.
  • Pack size: 10 g.

Catalog No. 50-002-09921 Supplier Medchemexpress LLC Supplier No. HYW05205110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzylacetamide is an acetamide derivative supplied for research use and reported as an endogenous metabolite and a major component of the plasma metabolic profile of benznidazole. It is provided as a white to off-white solid and is characterized by a molecular formula of C9H11NO and a molecular weight of 149.19 g·mol⁻¹. Storage recommendations are 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light).

  • Cas number: 588-46-5.
  • Chemical formula: C9H11NO.
  • Molecular weight: 149.19 g·mol⁻¹.
  • Purity: ≥97.0%.
  • Appearance: white to off-white solid.
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light).
  • Synonyms: N-acetylbenzylamine.
  • Pack size: 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.