Learn More
Medchemexpress LLC Acetamide, N-(phenylmethyl)- | 588-46-5 | MFCD00059204 | 99.4% | 149.19 g/mol | C9H11NO | 25 G
SDP
Catalog No. 5000209515
Encompass_Preferred
N-Benzylacetamide is a small-molecule endogenous metabolite (also called N-acetylbenzylamine) commonly used as a reference standard in metabolite profiling and analytical research. It is a white to off-white solid with molecular formula C9H11NO and a molecular weight of 149.19 g/mol.
  • Endogenous metabolite present in plasma metabolic profiles of benznidazole.
  • High purity suitable for analytical and profiling applications.
  • Available in multiple pack sizes, including 25 g, for scalability in experiments.
  • Provided with batch-specific datasheet and certificate of analysis for verification.
  • Store and handle per the SDS recommendations to maintain stability.
Catalog No. 50-002-09515 Supplier Medchemexpress LLC Supplier No. 25GHYW052051
N-Benzylacetamide is a small-molecule endogenous metabolite (also called N-acetylbenzylamine) commonly used as a reference standard in metabolite profiling and analytical research. It is a white to off-white solid with molecular formula C9H11NO and a molecular weight of 149.19 g/mol.
  • Endogenous metabolite present in plasma metabolic profiles of benznidazole.
  • High purity suitable for analytical and profiling applications.
  • Available in multiple pack sizes, including 25 g, for scalability in experiments.
  • Provided with batch-specific datasheet and certificate of analysis for verification.
  • Store and handle per the SDS recommendations to maintain stability.