Learn More
AdipoGen Acetamidofluorescein
SDP
Catalog No. 502561725
Encompass_Preferred
Chemical. CAS 82779-14-4. Formula C22H15NO6. MW 389.36. Acetamidofluorescein is a N-acetyl derivative of fluorescein that is strongly fluorescent und used as a fluorescent label. Spectral data lambdaex = 491nm, lambdaem 515nm.
Catalog No. 50-256-1725 Supplier AdipoGen Supplier No. CDXA0712M500
Chemical. CAS 82779-14-4. Formula C22H15NO6. MW 389.36. Acetamidofluorescein is a N-acetyl derivative of fluorescein that is strongly fluorescent und used as a fluorescent label. Spectral data lambdaex = 491nm, lambdaem 515nm.