Learn More
Medchemexpress LLC Acetaminophen | 103-90-2 | 99.97% | C8H9NO2 | 10 G
SDP
Catalog No. 5000434452
Encompass_Preferred
Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor and a widely used antipyretic and analgesic agent. It is also a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor that induces ferroptosis and can lead to acute liver injury in mice models.
  • Selective COX-2 inhibitor with an IC50 of 25.8 μM
  • Widely used as an antipyretic and analgesic
  • Potent hepatic N-acetyltransferase 2 (NAT2) inhibitor
  • Induces ferroptosis
  • Can lead to acute liver injury in mice models
Catalog No. 50-004-34452 Supplier Medchemexpress LLC Supplier No. HY6600510G
Acetaminophen (Paracetamol) is a selective cyclooxygenase-2 (COX-2) inhibitor and a widely used antipyretic and analgesic agent. It is also a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor that induces ferroptosis and can lead to acute liver injury in mice models.
  • Selective COX-2 inhibitor with an IC50 of 25.8 μM
  • Widely used as an antipyretic and analgesic
  • Potent hepatic N-acetyltransferase 2 (NAT2) inhibitor
  • Induces ferroptosis
  • Can lead to acute liver injury in mice models