Learn More
Medchemexpress LLC Acetaminophen | 103-90-2 | 99.97% | C8H9NO2 | 50 MG
SDP
Catalog No. 5000434728
Encompass_Preferred
Acetaminophen (Standard) is the analytical standard of Acetaminophen, intended for research and analytical applications. Also known as Paracetamol, it is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM, and a widely used antipyretic and analgesic agent. It also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Analytical standard grade
  • Reference standard supplied assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Suitable for HPLC
  • Suitable for GC
  • Suitable for MS
Catalog No. 50-004-34728 Supplier Medchemexpress LLC Supplier No. HY66005R50MG
Acetaminophen (Standard) is the analytical standard of Acetaminophen, intended for research and analytical applications. Also known as Paracetamol, it is a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM, and a widely used antipyretic and analgesic agent. It also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor.
  • Analytical standard grade
  • Reference standard supplied assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Suitable for HPLC
  • Suitable for GC
  • Suitable for MS