Learn More
U.S. Pharmacopeia Acetaminophen, 103-90-2, MFCD00002328, 400mg
Catalog No. NC9975429
Encompass
Molecular Formula C8H9NO2, Molecular Weight 151.164, Melting Point 334.4 - 341.6 °F (168 - 172 °C), Boiling Point > 932 °F (> 500 °C), Synonyms: Paracetamol, Acetamide, N-(4-hydroxyphenyl)-, 4'-hydroxyacetanilide
Catalog No. NC9975429 Supplier U.S. Pharmacopeia Supplier No. 1003009
Molecular Formula C8H9NO2, Molecular Weight 151.164, Melting Point 334.4 - 341.6 °F (168 - 172 °C), Boiling Point > 932 °F (> 500 °C), Synonyms: Paracetamol, Acetamide, N-(4-hydroxyphenyl)-, 4'-hydroxyacetanilide