Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Acetaminophen Serum Tox Calibrators

Catalog No. 23278398 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 5 mL, 5 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-398 1 x 5 mL, 5 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-398 Supplier Thermo Scientific™ Supplier No. 1091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use the Thermo Scientific™ DRI™ Acetaminophen Calibrator Set with the Thermo Scientific Acetaminophen Serum Toxicology Reagent Kit.

Use the Thermo Scientific™ DRI™ Acetaminophen Serum Toxicology Calibrator Set with the Thermo Scientific Acetaminophen Serum Toxicology Reagent Kit.

The DRI Acetaminophen Calibrator Kit contains 5mL of negative calibrator and 2mL each of 10µg/mL, 25µg/mL, 50µg/mL, 100µg/mL and 200µg/mL acetaminophen calibrators in Tris buffer with sodium azide as a preservative.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Acetaminophen Calibrator Set
Detectable Analytes Acetaminophen
Sample Type Serum
Type Calibrator
Level Negative, 10, 25, 50, 100, 200µg/mL
Quantity 1 x 5 mL, 5 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.